6-Ethoxy-2-naphthaleneboronic acid 98%

Warning

Synonyms: (6-Ethoxynaphthalen-2-yl)boronic acid

H52872-03 H52872-06
AAH52872-03EA 43.21 USD
AAH52872-03 AAH52872-06
6-Ethoxy-2-naphthaleneboronic acid 98%
6-Ethoxy-2-naphthaleneboronic acid
Formula: C₁₂H₁₃BO₃
MW: 216.04 g/mol
Melting Pt: 240…242 °C
Storage Temperature: Ambient
MDL Number: MFCD03427009
CAS Number: 352525-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR