3-(4-Morpholinylcarbonyl)phenylboronic acid 98%

Warning

Synonyms: 3-(Morpholine-4-carbonyl)phenylboronic acid3-(4-Morpholinylcarbonyl)benzeneboronic acid

H52868-03 H52868-MD
AAH52868-MDEA 87.24 USD
AAH52868-MD AAH52868-03
3-(4-Morpholinylcarbonyl)phenylboronic acid 98%
3-(4-Morpholinylcarbonyl)phenylboronic acid
Formula: C₁₁H₁₄BNO₄
MW: 235.05 g/mol
Melting Pt: 124…130 °C
Storage Temperature: Ambient
MDL Number: MFCD04115687
CAS Number: 723281-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR