3-Chloro-4-pyridineboronic acid 96%

Warning

H52864-03 H52864-MD
AAH52864-03EA 347.13 USD
AAH52864-03 AAH52864-MD
3-Chloro-4-pyridineboronic acid 96%
3-Chloro-4-pyridineboronic acid
Formula: C₅H₅BClNO₂
MW: 157.36 g/mol
Melting Pt: 95…97 °C
Storage Temperature: Ambient
MDL Number: MFCD03425942
CAS Number: 458532-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR