4-Chloro-2-ethoxyphenylboronic acid 98%

Warning

Synonyms: 4-Chloro-2-ethoxybenzeneboronic acid

H52861-03 H52861-MD
AAH52861-03EA 100.93 USD
AAH52861-03 AAH52861-MD
4-Chloro-2-ethoxyphenylboronic acid 98%
4-Chloro-2-ethoxyphenylboronic acid
Formula: C₈H₁₀BClO₃
MW: 200.43 g/mol
Melting Pt: 154…158 °C
Storage Temperature: Ambient
MDL Number: MFCD03094940
CAS Number: 850568-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR