Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(2-Pyridylcarbamoyl)phenylboronic acid 97%

Supplier: Alfa Aesar
Warning
Synonyms: 4-(2-Pyridylcarbamoyl)benzeneboronic acid

H52843-MD
AAH52843-MDEA 127.66 USD InStock
AAH52843-MD
4-(2-Pyridylcarbamoyl)phenylboronic acid 97%
4-(2-Pyridylcarbamoyl)phenylboronic acid


Formula: C₁₂H₁₁BN₂O₃
MW: 242,04
Melting Pt: 124-128°C
Storage Temperature: Ambient
MDL Number: MFCD06659876
CAS Number: 850568-25-1
EINECS: 000-000-0
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty