N-Ethyl 4-boronobenzenesulfonamide 97%

Warning

Synonyms: 4-(Ethylsulfamoyl)benzeneboronic acid4-(Ethylsulphamoyl)benzeneboronic acid

H52840-03 H52840-MD
AAH52840-MDEA 95.68 USD
AAH52840-MD AAH52840-03
N-Ethyl 4-boronobenzenesulfonamide 97%
N-Ethyl 4-boronobenzenesulphonamide
Formula: C₈H₁₂BNO₄S
MW: 229.06 g/mol
Melting Pt: 132…136 °C
Storage Temperature: Ambient
MDL Number: MFCD07363751
CAS Number: 871329-65-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR