(S)-3-(Aminomethyl)-1-boc-piperidine 97%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: (S)-1-BOC-3-(Aminomethyl)piperidine

H52822-MD H52822-06 H52822-03
AAH52822-MDEA 50.6 USD
AAH52822-MD AAH52822-06 AAH52822-03
(S)-3-(Aminomethyl)-1-boc-piperidine 97%
(S)-3-(Aminomethyl)-1-Boc-piperidine
Formula: C₁₁H₂₂N₂O₂
MW: 214.31
Storage Temperature: Ambient
MDL Number: MFCD03839877
CAS Number: 140645-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR