(R)-2-Hydroxy-4-phenylbutyric acid 97%

Supplier: Thermo Scientific Chemicals

H52804-06 H52804-03
AAH52804-03EA 39.99 USD
AAH52804-03 AAH52804-06
(R)-2-Hydroxy-4-phenylbutyric acid 97%
(R)-2-Hydroxy-4-phenylbutyric acid
Formula: C₁₀H₁₂O₃
MW: 180.20 g/mol
Melting Pt: 114…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00192219
CAS Number: 29678-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR