2-(Trifluoromethyl)pyrimidine-4-carboxamide 97%

Supplier: Thermo Scientific Chemicals
Warning

H52803-03 H52803-MD
AAH52803-MDEA 119.62 USD
AAH52803-MD AAH52803-03
2-(Trifluoromethyl)pyrimidine-4-carboxamide 97%
2-(Trifluoromethyl)pyrimidine-4-carboxamide
Formula: C₆H₄F₃N₃O
Storage Temperature: Ambient
MDL Number: MFCD05662689
CAS Number: 914348-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR