1-Boc-2-methyl-L-proline 97%

Synonyms: (S)-1-(tert-Butoxycarbonyl)-2-methylpyrrolidine-2-carboxylic acid

H52796-MD H52796-03
AAH52796-03EA 597.18 USD
AAH52796-03 AAH52796-MD
1-Boc-2-methyl-L-proline 97%
1-Boc-2-methyl-L-proline
Formula: C₁₁H₁₉NO₄
MW: 229.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08706361
CAS Number: 103336-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR