(±)-3-(Boc-amino)piperidine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Aminopiperidine, 3-BOC protected
, tert-Butyl piperidin-3-ylcarbamate
H52788-06
H52788-03
AAH52788-06EA
196
USD
AAH52788-06
AAH52788-03
(±)-3-(Boc-amino)piperidine 97%
(±)-3-(Boc-amino)piperidine
Formula:
C₁₀H₂₀N₂O₂ MW: 200.28 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03839941 CAS Number: 172603-05-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...