3-(Isopropoxycarbonyl)phenylboronic acid 98%

Warning

Synonyms: 3-(Isopropoxycarbonyl)benzeneboronic acid

H52758-MD H52758-03
AAH52758-03EA 142.4 USD
AAH52758-03 AAH52758-MD
3-(Isopropoxycarbonyl)phenylboronic acid 98%
3-(Isopropoxycarbonyl)phenylboronic acid
Formula: C₁₀H₁₃BO₄
MW: 208.02
Melting Pt: 127…129 °C
Storage Temperature: Ambient
MDL Number: MFCD02093047
CAS Number: 342002-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR