5-Fluoro-(2-methoxycarbonyl)phenylboronic acid 97%

Warning

Synonyms: 2-Methoxycarbonyl-5-fluorophenylboronic acid5-Fluoro-2-(methoxycarbonyl)benzeneboronic acid

H52756-03 H52756-MD
AAH52756-03EA 167.08 USD
AAH52756-03 AAH52756-MD
5-Fluoro-(2-methoxycarbonyl)phenylboronic acid 97%
5-Fluoro-(2-methoxycarbonyl)phenylboronic acid
Formula: C₈H₈BFO₄
MW: 197.96 g/mol
Melting Pt: 146…148 °C
Storage Temperature: Ambient
MDL Number: MFCD03095367
CAS Number: 850568-05-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR