3-Fluoro-2-methylphenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (3-Fluoro-2-methylphenyl)boronic acid3-Fluoro-2-methylbenzeneboronic acid

H52752-03 H52752-MD
AAH52752-03EA 227.99 USD
AAH52752-03 AAH52752-MD
3-Fluoro-2-methylphenylboronic acid 98%
3-Fluoro-2-methylphenylboronic acid
Formula: C₇H₈BFO₂
MW: 153.95 g/mol
Melting Pt: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD07368679
CAS Number: 163517-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR