(R)-3-Hydroxypyrrolidine hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (3R)-pyrrolidin-3-ol hydrochloride(R)-3-Hydroxypyrrolidine HCl(R)-3-Pyrrolidinol hydrochloride

H52743-03 H52743-06
AAH52743-03EA 26.88 USD
AAH52743-03 AAH52743-06
(R)-3-Hydroxypyrrolidine hydrochloride 97%
(R)-3-Hydroxypyrrolidine hydrochloride
Formula: C₄H₁₀ClNO
MW: 123.58 g/mol
Melting Pt: 104…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00191570
CAS Number: 104706-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR