3-Chloro-4-(methylcarbamoyl)phenylboronic acid 96%

Warning

Synonyms: 3-Chloro-4-(methylcarbamoyl)benzeneboronic acid(3-Chloro-4-(methylcarbamoyl)phenyl)boronic acid

H52721-MD H52721-03
AAH52721-03EA 325.23 USD
AAH52721-03 AAH52721-MD
3-Chloro-4-(methylcarbamoyl)phenylboronic acid 96%
3-Chloro-4-(methylcarbamoyl)phenylboronic acid
Formula: C₈H₉BClNO₃
MW: 213.43 g/mol
Melting Pt: 164…170 °C
Storage Temperature: Ambient
MDL Number: MFCD07363766
CAS Number: 850589-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR