3-Benzyloxy-5-fluorophenylboronic acid 98%

Warning

Synonyms: 3-Benzyloxy-5-fluorobenzeneboronic acid

H52704-03 H52704-MD
AAH52704-03EA 163.17 USD
AAH52704-03 AAH52704-MD
3-Benzyloxy-5-fluorophenylboronic acid 98%
3-Benzyloxy-5-fluorophenylboronic acid
Formula: C₁₃H₁₂BFO₃
MW: 246.04 g/mol
Melting Pt: 156…162 °C
Storage Temperature: Ambient
MDL Number: MFCD07363782
CAS Number: 850589-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR