3-(Ethylcarbamoyl)phenylboronic acid 97%

Warning

Synonyms: 3-(Ethylcarbamoyl)benzeneboronic acid3-(N-Ethylaminocarbonyl)phenylboronic acid

H52696-03 H52696-MD
AAH52696-MDEA 78.62 USD
AAH52696-MD AAH52696-03
3-(Ethylcarbamoyl)phenylboronic acid 97%
3-(Ethylcarbamoyl)phenylboronic acid
Formula: C₉H₁₂BNO₃
MW: 193.01 g/mol
Melting Pt: 106…116 °C
Storage Temperature: Ambient
MDL Number: MFCD04115689
CAS Number: 850567-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR