3-(Butylcarbamoyl)phenylboronic acid 97%

Warning

Synonyms: 3-(n-Butylcarbamoyl)benzeneboronic acid(3-(Butylcarbamoyl)phenyl)boronic acid

H52693-03 H52693-MD
AAH52693-MDEA 47.26 USD
AAH52693-MD AAH52693-03
3-(Butylcarbamoyl)phenylboronic acid 97%
3-(Butylcarbamoyl)phenylboronic acid
Formula: C₁₁H₁₆BNO₃
MW: 221.06 g/mol
Melting Pt: 216…224 °C
Storage Temperature: Ambient
MDL Number: MFCD04115691
CAS Number: 397843-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR