3-Chloro-4-(piperidine-1-carbonyl)phenylboronic acid 98%

Warning

Synonyms: 3-Chloro-4-(1-piperidinylcarbonyl)benzeneboronic acid

H52685-03 H52685-MD
AAH52685-MDEA 100.93 USD
AAH52685-MD AAH52685-03
3-Chloro-4-(piperidine-1-carbonyl)phenylboronic acid 98%
3-Chloro-4-(piperidine-1-carbonyl)phenylboronic acid
Formula: C₁₂H₁₅BClNO₃
MW: 267.52
Melting Pt: 160…163 °C
Storage Temperature: Ambient
MDL Number: MFCD07363775
CAS Number: 850589-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR