2-(trans-2-Cyanovinyl)benzeneboronic acid 95%

Warning

H52652-MD H52652-03
AAH52652-MDEA 103.92 USD
AAH52652-MD AAH52652-03
2-(trans-2-Cyanovinyl)benzeneboronic acid 95%
2-(trans-2-Cyanovinyl)benzeneboronic acid
Formula: C₉H₈BNO₂
MW: 172.98
Storage Temperature: Ambient
MDL Number: MFCD06659825
CAS Number: 850568-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR