4-Fluoro-3-methoxyphenylboronic acid 98%

Warning

Synonyms: 4-Fluoro-3-methoxybenzeneboronic acid(4-Fluoro-3-methoxyphenyl)boronic acid

H52635-MD H52635-03
AAH52635-03EA 252.16 USD
AAH52635-03 AAH52635-MD
4-Fluoro-3-methoxyphenylboronic acid 98%
4-Fluoro-3-methoxyphenylboronic acid
Formula: C₇H₈BFO₃
MW: 169.95 g/mol
Melting Pt: 189…191 °C
Storage Temperature: Ambient
MDL Number: MFCD08056358
CAS Number: 854778-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR