4-[5-(4-(Dimethylamino)phenyl)oxazol-2-yl]phenylboronic acid 97%

Warning

Synonyms: 4-[5-(4-Dimethylaminophenyl)-2-oxazolyl]benzeneboronic acid

H52632-MD
AAH52632-MDEA 634.92 USD
AAH52632-MD
4-[5-(4-(Dimethylamino)phenyl)oxazol-2-yl]phenylboronic acid 97%
4-[5-(4-(Dimethylamino)phenyl)oxazol-2-yl]phenylboronic acid
Formula: C₁₇H₁₇BN₂O₃
MW: 308.14
Storage Temperature: Ambient
MDL Number: MFCD08689479
CAS Number: 380499-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR