4-(Benzyloxy)phenylboronic acid pinacol ester 98%

Warning

Synonyms: 2-(4-(Benzyloxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane4-Benzyloxybenzeneboronic acid pinacol ester

H52630-03 H52630-06
AAH52630-06EA 294.77 USD
AAH52630-06 AAH52630-03
4-(Benzyloxy)phenylboronic acid pinacol ester 98%
4-(Benzyloxy)phenylboronic acid pinacol ester
Formula: C₁₉H₂₃BO₃
MW: 310.19 g/mol
Melting Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD04974107
CAS Number: 754226-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR