3-Boronobenzenesulfonamide 97%

Warning

Synonyms: 3-(Aminosulfonyl)benzeneboronic acid3-Sulfamoylbenzeneboronic acid3-(Aminosulphonyl)benzeneboronic acid3-Sulphamoylbenzeneboronic acid

H52628-MD
AAH52628-MDEA 458.43 USD
AAH52628-MD
3-Boronobenzenesulfonamide 97%
3-Boronobenzenesulphonamide
Formula: C₆H₈BNO₄S
MW: 201.00 g/mol
Melting Pt: 260…266 °C
Storage Temperature: Ambient
MDL Number: MFCD06659840
CAS Number: 850568-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR