3-(Cyanomethyl)benzeneboronic acid pinacol ester 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetonitrile2-[3-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile

H52618-MD H52618-06 H52618-03
AAH52618-MDEA 33.72 USD
AAH52618-MD AAH52618-06 AAH52618-03
3-(Cyanomethyl)benzeneboronic acid pinacol ester 96%
3-(Cyanomethyl)benzeneboronic acid pinacol ester
Formula: C₁₄H₁₈BNO₂
MW: 243.113 g/mol
Melting Pt: 44…48 °C
Storage Temperature: Ambient
MDL Number: MFCD02179458
CAS Number: 396131-82-1

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR