3,5-Dinitro-2-methylphenylboronic acid 96%

Warning

Synonyms: 3,5-Dinitro-2-methylbenzeneboronic acid

H52611-03 H52611-06
AAH52611-03EA 35.77 USD
AAH52611-03 AAH52611-06
3,5-Dinitro-2-methylphenylboronic acid 96%
3,5-Dinitro-2-methylphenylboronic acid
Formula: C₇H₇BN₂O₆
MW: 225.95
Melting Pt: 122…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00757434
CAS Number: 24341-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR