4-(2-Hydroxyethylcarbamoyl)phenylboronic acid 98%

Warning

Synonyms: 4-(2-Hydroxyethylcarbamoyl)benzeneboronic acid

H52608-03 H52608-MD
AAH52608-03EA 233.1 USD
AAH52608-03 AAH52608-MD
4-(2-Hydroxyethylcarbamoyl)phenylboronic acid 98%
4-(2-Hydroxyethylcarbamoyl)phenylboronic acid
Formula: C₉H₁₂BNO₄
MW: 209.01
Melting Pt: 228…230 °C
Storage Temperature: Ambient
MDL Number: MFCD06659848
CAS Number: 850593-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR