a-Tosyl(4-fluorobenzyl)isocyanide 97%

Warning

Synonyms: (4-Fluorophenyl)(isocyano)methyl 4-methylphenyl sulphonea-Tosyl-(4-fluorobenzyl) isocyanide

H52580-MD H52580-03
AAH52580-03EA 256.83 USD
AAH52580-03 AAH52580-MD
a-Tosyl(4-fluorobenzyl)isocyanide 97%
a-Tosyl(4-fluorobenzyl)isocyanide
Formula: C₁₅H₁₂FNO₂S
MW: 289.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03787932
CAS Number: 165806-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR