3-(Dimethylcarbamoyl)phenylboronic acid 96%

Warning

Synonyms: 3-(Dimethylcarbamoyl)benzeneboronic acid(3-(Dimethylcarbamoyl)phenyl)boronic acid

H52545-MD H52545-03
AAH52545-MDEA 40.46 USD
AAH52545-MD AAH52545-03
3-(Dimethylcarbamoyl)phenylboronic acid 96%
3-(Dimethylcarbamoyl)phenylboronic acid
Formula: C₉H₁₂BNO₃
MW: 193.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03412066
CAS Number: 373384-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR