2-Ethoxy-5-(trifluoromethyl)phenylboronic acid 98%

Warning

Synonyms: 2-Ethoxy-5-trifluoromethylphenylboronic acid2-Ethoxy-5-(trifluoromethyl)benzeneboronic acid

H52541-MD H52541-03
AAH52541-03EA 136.04 USD
AAH52541-03 AAH52541-MD
2-Ethoxy-5-(trifluoromethyl)phenylboronic acid 98%
2-Ethoxy-5-(trifluoromethyl)phenylboronic acid
Formula: C₉H₁₀BF₃O₃
MW: 233.98 g/mol
Melting Pt: 100…104 °C
Storage Temperature: Ambient
MDL Number: MFCD06801695
CAS Number: 850593-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR