Ethyl-3-(3-boronobenzamido)propionate 97%

Warning

Synonyms: (3-((3-Ethoxy-3-oxopropyl)carbamoyl)phenyl)boronic acid3-[(3-Ethoxy-3-oxopropyl)carbamoyl]benzeneboronic acid

H52539-MD H52539-03
AAH52539-MDEA 81.27 USD
AAH52539-MD AAH52539-03
Ethyl-3-(3-boronobenzamido)propionate 97%
Ethyl 3-(3-boronobenzamido)propionate
Formula: C₁₂H₁₆BNO₅
MW: 265.07 g/mol
Melting Pt: 128…132 °C
Storage Temperature: Ambient
MDL Number: MFCD04115701
CAS Number: 850567-28-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR