4-Chloro-2-methoxyphenylboronic acid 98%

Warning

Synonyms: (4-Chloro-2-methoxyphenyl)boronic acid4-Chloro-2-methoxybenzeneboronic acid

H52529-03 H52529-MD
AAH52529-03EA 247.45 USD
AAH52529-03 AAH52529-MD
4-Chloro-2-methoxyphenylboronic acid 98%
4-Chloro-2-methoxyphenylboronic acid
Formula: C₇H₈BClO₃
MW: 186.40 g/mol
Melting Pt: 128…134 °C
Storage Temperature: Ambient
MDL Number: MFCD02684317
CAS Number: 762287-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR