3-Pyridineboronic acid pinacol ester 98%

Warning

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridinePyridine-3-boronic acid pinacol ester

H52513-MD H52513-03
AAH52513-MDEA 53.75 USD
AAH52513-MD AAH52513-03
3-Pyridineboronic acid pinacol ester 98%
3-Pyridineboronic acid pinacol ester
Formula: C₁₁H₁₆BNO₂
MW: 205.06 g/mol
Melting Pt: 77…80 °C
Storage Temperature: Ambient
MDL Number: MFCD03084758
CAS Number: 329214-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR