N,N-Diethyl 3-borono-4-methylbenzenesulfonamide 97%

Warning

Synonyms: (5-(N,N-Diethylsulphamoyl)-2-methylphenyl)boronic acid5-Diethylsulfamoyl-2-methylbenzeneboronic acid5-Diethylsulphamoyl-2-methylbenzeneboronic acid(5-(N,N-Diethylsulfamoyl)-2-methylphenyl)boronic acid

H52509-03 H52509-MD
AAH52509-MDEA 69.2 USD
AAH52509-MD AAH52509-03
N,N-Diethyl 3-borono-4-methylbenzenesulfonamide 97%
N,N-Diethyl 3-borono-4-methylbenzenesulphonamide
Formula: C₁₁H₁₈BNO₄S
MW: 271.14 g/mol
Melting Pt: 146…152 °C
Storage Temperature: Ambient
MDL Number: MFCD08056356
CAS Number: 871329-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR