2-Phenylethylboronic acid 98%

Warning

Synonyms: Phenethylboronic acid

H52502-03 H52502-06
AAH52502-03EA 38.32 USD
AAH52502-03 AAH52502-06
2-Phenylethylboronic acid 98%
2-Phenylethylboronic acid
Formula: C₈H₁₁BO₂
MW: 149.98 g/mol
Melting Pt: 82…84 °C
Storage Temperature: Ambient
MDL Number: MFCD01631226
CAS Number: 34420-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR