5-Bromo-2-methoxyphenylboronic acid 97%
|
Warning
|
Synonyms:
5-Bromo-2-methoxybenzeneboronic acid
H52497-03
H52497-06
AAH52497-06EA
96.86
USD
AAH52497-06
AAH52497-03
5-Bromo-2-methoxyphenylboronic acid 97%
5-Bromo-2-methoxyphenylboronic acid
Formula:
C₇H₈BBrO₃ MW: 230.85 g/mol Melting Pt: 129…131 °C Storage Temperature: Ambient |
MDL Number:
MFCD00093409 CAS Number: 89694-45-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...