N-Methyl 4-boronobenzenesulfonamide 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Methylsulphamoylbenzeneboronic acid(4-(N-Methylsulphamoyl)phenyl)boronic acid(4-(N-Methylsulfamoyl)phenyl)boronic acid4-Methylsulfamoylbenzeneboronic acid

H52493-MD H52493-03
AAH52493-03EA 384.99 USD
AAH52493-03 AAH52493-MD
N-Methyl 4-boronobenzenesulfonamide 97%
N-Methyl 4-boronobenzenesulphonamide
Formula: C₇H₁₀BNO₄S
MW: 215.03 g/mol
Melting Pt: 136…140 °C
Storage Temperature: Ambient
MDL Number: MFCD06659864
CAS Number: 226396-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR