4-Carboxy-2-chlorophenylboronic acid 97%

Warning

Synonyms: 4-Carboxy-2-chlorobenzeneboronic acid

H52489-06 H52489-03
AAH52489-06EA 296.37 USD
AAH52489-06 AAH52489-03
4-Carboxy-2-chlorophenylboronic acid 97%
4-Carboxy-2-chlorophenylboronic acid
Formula: C₇H₆BClO₄
MW: 200.39 g/mol
Melting Pt: 208…210 °C
Storage Temperature: Ambient
MDL Number: MFCD07363763
CAS Number: 851335-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR