4-Fluoro-3-(trifluoromethyl)phenylboronic acid 98%

Warning

Synonyms: 4-Fluoro-3-(trifluoromethyl)benzeneboronic acid

H52483-06 H52483-03
AAH52483-03EA 58.82 USD
AAH52483-03 AAH52483-06
4-Fluoro-3-(trifluoromethyl)phenylboronic acid 98%
4-Fluoro-3-(trifluoromethyl)phenylboronic acid
Formula: C₇H₅BF₄O₂
MW: 207.92 g/mol
Melting Pt: 170…174 °C
Storage Temperature: Ambient
MDL Number: MFCD07363787
CAS Number: 182344-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR