2-(2,2,2-Trifluoroethoxy)phenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2-(2,2,2-Trifluoroethoxy)phenyl)boronic acid2-(2,2,2-Trifluoroethoxy)benzeneboronic acid

H52471-03 H52471-MD
AAH52471-03EA 411.81 USD
AAH52471-03 AAH52471-MD
2-(2,2,2-Trifluoroethoxy)phenylboronic acid 98%
2-(2,2,2-Trifluoroethoxy)phenylboronic acid
Formula: C₈H₈BF₃O₃
MW: 219.95 g/mol
Melting Pt: 125…128 °C
Storage Temperature: Ambient
MDL Number: MFCD09027258
CAS Number: 957060-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR