(4-Benzamidophenyl)boronic acid 95%

Warning

Synonyms: 4-Benzamidobenzeneboronic acid

H52456-MD H52456-03
AAH52456-MDEA 102.56 USD
AAH52456-MD AAH52456-03
(4-Benzamidophenyl)boronic acid 95%
(4-Benzamidophenyl)boronic acid
Formula: C₁₃H₁₂BNO₃
MW: 241.05 g/mol
Melting Pt: 194…196 °C
Storage Temperature: Ambient
MDL Number: MFCD08235089
CAS Number: 397843-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR