2-(Hydroxymethyl)phenylboronic acid 97%
|
Warning
|
Synonyms:
2-(Hydroxymethyl)benzeneboronic acid
, (2-(Hydroxymethyl)phenyl)boronic acid
H52442-06
H52442-03
AAH52442-06EA
407.88
USD
AAH52442-06
AAH52442-03
2-(Hydroxymethyl)phenylboronic acid 97%
2-(Hydroxymethyl)phenylboronic acid
Formula:
C₇H₉BO₃ MW: 151.96 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02258966 CAS Number: 87199-14-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...