N-Cyclopropyl 3-boronobenzenesulfonamide 98%

Warning

Synonyms: (3-(N-Cyclopropylsulphamoyl)phenyl)boronic acid3-(Cyclopropylsulfamoyl)benzeneboronic acid3-(Cyclopropylsulphamoyl)benzeneboronic acid(3-(N-Cyclopropylsulfamoyl)phenyl)boronic acid

H52441-MD H52441-03
AAH52441-MDEA 136.53 USD
AAH52441-MD AAH52441-03
N-Cyclopropyl 3-boronobenzenesulfonamide 98%
N-Cyclopropyl 3-boronobenzenesulphonamide
Formula: C₉H₁₂BNO₄S
MW: 241.07 g/mol
Melting Pt: 126…128 °C
Storage Temperature: Ambient
MDL Number: MFCD08436035
CAS Number: 913835-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR