Methyl-3-(2-bromoethoxy)benzenecarboxylate ≥97%

Warning

H52405-06 H52405-MD H52405-03
AAH52405-MDEA 52.07 USD
AAH52405-MD AAH52405-03 AAH52405-06
Methyl-3-(2-bromoethoxy)benzenecarboxylate ≥97%
Methyl-3-(2-bromoethoxy)benzenecarboxylate
Formula: C₁₀H₁₁BrO₃
MW: 259.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06204041
CAS Number: 59516-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR