2-Methoxynicotinonitrile 97+%

Warning

Synonyms: 3-Cyano-2-methoxypyridine

H52373-MD H52373-06 H52373-03
AAH52373-03EA 129 USD
AAH52373-03 AAH52373-MD AAH52373-06
2-Methoxynicotinonitrile 97+%
2-Methoxynicotinonitrile 2-(4-Formylphenyl)isonicotinonitrile
Formula: C₇H₆N₂O
MW: 134.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07363804
CAS Number: 7254-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR