2,4,5-Trimethyl-3-pyridinol 97%

Supplier: Thermo Scientific Chemicals
Warning

H52357-MD H52357-03
AAH52357-MDEA 81.39 USD
AAH52357-MD AAH52357-03
2,4,5-Trimethyl-3-pyridinol 97%
2,4,5-Trimethyl-3-pyridinol
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Melting Pt: 172…176 °C
Storage Temperature: Ambient
MDL Number: MFCD17015035
CAS Number: 5622-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR