2,6-Dichloro-4-methylnicotinic acid 98%

Warning

H52347-MD H52347-03 H52347-06
AAH52347-03EA 137.23 USD
AAH52347-03 AAH52347-MD AAH52347-06
2,6-Dichloro-4-methylnicotinic acid 98%
2,6-Dichloro-4-methylnicotinic acid
Formula: C₇H₅Cl₂NO₂
MW: 206.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02932724
CAS Number: 62774-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR