2-Bromo-6-fluorophenylacetonitrile 96%

Warning

H52341-MD H52341-03
AAH52341-03EA 338.94 USD
AAH52341-03 AAH52341-MD
2-Bromo-6-fluorophenylacetonitrile 96%
2-Bromo-6-fluorophenylacetonitrile
Formula: C₈H₅BrFN
MW: 214.04 g/mol
Melting Pt: 60…62 °C
Storage Temperature: Ambient
MDL Number: MFCD11226884
CAS Number: 936693-22-0
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR