4-Indoleboronic acid pinacol ester 97+%

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

H52325-06 H52325-03 H52325-MD
AAH52325-03EA 147.85 USD
AAH52325-03 AAH52325-06 AAH52325-MD
4-Indoleboronic acid pinacol ester 97+%
4-Indoleboronic acid pinacol ester
Formula: C₁₄H₁₈BNO₂
MW: 243.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08689896
CAS Number: 388116-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR